Learn More
Medchemexpress LLC SKI-I 25mg | 306301-68-8 | 25 MG
SDP
Catalog No. 5000307707
Encompass_Preferred
SKI-I is a research-grade small-molecule inhibitor of sphingosine kinase used in biochemical and cell-based assays to study sphingolipid signaling and apoptosis. Not for human use.
  • Potent sphingosine kinase inhibitor (IC50 1.2 μM for ST-hSK).
  • Also inhibits hERK2 (IC50 11 μM).
  • Induces apoptosis in multiple tumor cell lines.
  • High purity (99.14%).
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
Catalog No. 50-003-07707 Supplier Medchemexpress LLC Supplier No. HY11573525MG
SKI-I is a research-grade small-molecule inhibitor of sphingosine kinase used in biochemical and cell-based assays to study sphingolipid signaling and apoptosis. Not for human use.
  • Potent sphingosine kinase inhibitor (IC50 1.2 μM for ST-hSK).
  • Also inhibits hERK2 (IC50 11 μM).
  • Induces apoptosis in multiple tumor cell lines.
  • High purity (99.14%).
  • Soluble in DMSO at 100 mg/mL for in vitro applications.