Learn More
Medchemexpress LLC SKI-I | 5MG
SDP
Catalog No. 5000228050
Encompass_Preferred
SKI-I is a potent and selective inhibitor of human sphingosine kinase (SK) with an IC50 of 1 2 M for ST-hSK SKI-I also inhibits hERK2 (IC50 11 M) SKI-I induces apoptosis in tumor cell lines[1][2]
Catalog No. 50-002-28050 Supplier Medchemexpress LLC Supplier No. HY1157355MG
SKI-I is a potent and selective inhibitor of human sphingosine kinase (SK) with an IC50 of 1 2 M for ST-hSK SKI-I also inhibits hERK2 (IC50 11 M) SKI-I induces apoptosis in tumor cell lines[1][2]