Learn More
Medchemexpress LLC 4-((4-(4-chlorophenyl)-2-thiazolyl)amino)phenol | 312636-16-1 | MFCD00733553 | 99.3% | C15H11ClN2OS | 10MG
SDP
Catalog No. 502030482
Encompass_Preferred
SKI II is a small-molecule research inhibitor of sphingosine kinase used to probe SK1 and SK2 activity in biochemical and cellular studies. It is supplied as a white to off-white solid with high purity and is intended for research use only.
  • High purity (~99.3% by LCMS) suitable for analytical and biological experiments.
  • Inhibits sphingosine kinase with reported IC50 values of ~78 μM (SK1) and ~45 μM (SK2).
  • Cell-permeable, enabling use in cell-based assays.
  • Stable when stored as recommended: powder at -20°C, in solvent at -80°C.
  • Provided with certificate of analysis and safety data for quality and handling information.
Catalog No. 50-203-0482 Supplier Medchemexpress LLC Supplier No. HY1382210MG
SKI II is a small-molecule research inhibitor of sphingosine kinase used to probe SK1 and SK2 activity in biochemical and cellular studies. It is supplied as a white to off-white solid with high purity and is intended for research use only.
  • High purity (~99.3% by LCMS) suitable for analytical and biological experiments.
  • Inhibits sphingosine kinase with reported IC50 values of ~78 μM (SK1) and ~45 μM (SK2).
  • Cell-permeable, enabling use in cell-based assays.
  • Stable when stored as recommended: powder at -20°C, in solvent at -80°C.
  • Provided with certificate of analysis and safety data for quality and handling information.