Learn More
Selleck Chemical LLC SKI II
SDP
Supplier:  Selleck Chemical LLC S717610MM/1ML
Encompass_Preferred
SKI II (SphK-I2) is a highly selective and non ATP-competitive sphingosine kinase (SphK) inhibitor with IC50 of 0 5 M while exhibiting no inhibitory action on other kinases including PI3K PKC and ERK2
Catalog No. 50-000-04756