Learn More
Apexbio Technology LLC SKI II 312636-16-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061556
Encompass_Preferred
SKI II (CAS 312636-16-1) is a small-molecule inhibitor of sphingosine kinase (SK) an enzyme involved in converting sphingosine into sphingosine-1-phosphate (S1P) SKI II inhibits SK activity with an IC50 of 0 5 M through a mechanism independent of competition at the ATP-binding site Selectivity assessments showed no inhibitory effect against human kinases ERK2 PKC-I or PI3K Cellular assays demonstrate that SKI II effectively inhibits endogenous SK reducing S1P synthesis and subsequently suppressing proliferation and inducing apoptosis in various cancer cell lines including MDA-MB-231 T-24 MCF-7 and MCF-7/VP cells
Catalog No. 50-000-61556 Supplier Apexbio Technology LLC Supplier No. B22265.1
SKI II (CAS 312636-16-1) is a small-molecule inhibitor of sphingosine kinase (SK) an enzyme involved in converting sphingosine into sphingosine-1-phosphate (S1P) SKI II inhibits SK activity with an IC50 of 0 5 M through a mechanism independent of competition at the ATP-binding site Selectivity assessments showed no inhibitory effect against human kinases ERK2 PKC-I or PI3K Cellular assays demonstrate that SKI II effectively inhibits endogenous SK reducing S1P synthesis and subsequently suppressing proliferation and inducing apoptosis in various cancer cell lines including MDA-MB-231 T-24 MCF-7 and MCF-7/VP cells