Learn More
Abcam SKI-II, Sphingosine kinase (SK1/2) inhibitor, 10MG
Catalog No. 502706501
Encompass_Preferred
MW 302.8 Da, Purity >98%. Sphingosine kinase (SK1/2) inhibitor (IC₅₀ values are 35 and 20 μM for SK1 and SK2 respectively). Inhibits tumor growth in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6501 Supplier Abcam Supplier No. AB14159410MG
MW 302.8 Da, Purity >98%. Sphingosine kinase (SK1/2) inhibitor (IC₅₀ values are 35 and 20 μM for SK1 and SK2 respectively). Inhibits tumor growth in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement