Learn More
Medchemexpress LLC SKI V | 10 MG
SDP
Catalog No. 5000339649
Encompass_Preferred
SKI V is a noncompetitive and potent non-lipid sphingosine kinase (SPHK; SK) inhibitor with an IC50 of 2 μM for GST-hSK. It potently inhibits PI3K with an IC50 of 6 μM for hPI3k. SKI V decreases the formation of the mitogenic second messenger sphingosine-1-phosphate (S1P), induces apoptosis, and exhibits antitumor activity.
  • Noncompetitive and potent non-lipid sphingosine kinase (SPHK; SK) inhibitor.
  • Inhibits PI3K.
  • Decreases formation of sphingosine-1-phosphate (S1P).
  • Induces apoptosis.
  • Possesses antitumor activity.
Catalog No. 50-003-39649 Supplier Medchemexpress LLC Supplier No. HY1289510MG
SKI V is a noncompetitive and potent non-lipid sphingosine kinase (SPHK; SK) inhibitor with an IC50 of 2 μM for GST-hSK. It potently inhibits PI3K with an IC50 of 6 μM for hPI3k. SKI V decreases the formation of the mitogenic second messenger sphingosine-1-phosphate (S1P), induces apoptosis, and exhibits antitumor activity.
  • Noncompetitive and potent non-lipid sphingosine kinase (SPHK; SK) inhibitor.
  • Inhibits PI3K.
  • Decreases formation of sphingosine-1-phosphate (S1P).
  • Induces apoptosis.
  • Possesses antitumor activity.