Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ski V | 24418-86-8 | MFCD07772193 | 99.0% | 254.24 g/mol | C15H10O4 | 5 MG
SDP

Catalog No. 5000263008
Encompass_Preferred

SKI V is a small-molecule sphingosine kinase inhibitor reported to induce apoptosis and exhibit antitumor activity in vitro. The compound is supplied as a powder for laboratory research, with molecular formula C15H10O4 and molecular weight 254.24 g/mol. Solubility and activity data support formulation in DMSO and mixed aqueous vehicles for cell-based studies.

  • Inhibits sphingosine kinase and PI3K signaling.
  • Demonstrates in vitro activity (IC50 ≈ 2 μM for GST-hSK).
  • Reported IC50 values for additional targets provide selectivity context.
  • Supplied as a powder suitable for laboratory handling and storage.
  • Soluble in DMSO (~50 mg/mL) and formulatable in aqueous vehicles with co-solvents.
  • Molecular weight 254.24 g/mol; chemical formula C15H10O4.
  • Intended for research use only.

Catalog No. 50-002-63008 Supplier Medchemexpress LLC Supplier No. HY128955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKI V is a small-molecule sphingosine kinase inhibitor reported to induce apoptosis and exhibit antitumor activity in vitro. The compound is supplied as a powder for laboratory research, with molecular formula C15H10O4 and molecular weight 254.24 g/mol. Solubility and activity data support formulation in DMSO and mixed aqueous vehicles for cell-based studies.

  • Inhibits sphingosine kinase and PI3K signaling.
  • Demonstrates in vitro activity (IC50 ≈ 2 μM for GST-hSK).
  • Reported IC50 values for additional targets provide selectivity context.
  • Supplied as a powder suitable for laboratory handling and storage.
  • Soluble in DMSO (~50 mg/mL) and formulatable in aqueous vehicles with co-solvents.
  • Molecular weight 254.24 g/mol; chemical formula C15H10O4.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.