Learn More
Aviva Systems Biology Corp SKIIP Antibody - N-terminal region (ARP32294_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32294_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SKIIP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SKIIP. Alternate names: Bx42, SKIP, Prp45, SKIIP, PRPF45, NCOA-62ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-54514