Learn More
Aviva Systems Biology Corp SKIL Antibody - N-terminal region (ARP32371_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32371_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SKIL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SKIL. Alternate names: SNO, SnoA, SnoI, SnoNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54549