Learn More
Aviva Systems Biology Corp SKIL Antibody - middle region (ARP58074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SKIL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human SKIL. Alternate names: SNO, SnoA, SnoI, SnoNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-435