Learn More
Enterprise Technology Solutions SKILCRAFT Calcium Lime Remover, Biobased, 5 Gallon, Green
SDP
Catalog No. 5000790244
Encompass_Preferred
Calcium Lime Remover (CLR) is a powerful formula that eliminates built-up mineral deposits and eradicates rust stains. It is perfect for use on tubs, shower heads, glassware, sinks, fiberglass, humidifiers, dishwashers and more. USDA Biopreferred Program Certified remover comes in a convenient bucket with a wire handle.
Catalog No. 50-007-90244 Supplier Enterprise Technology Solutions Supplier No. NSN5606131
Calcium Lime Remover (CLR) is a powerful formula that eliminates built-up mineral deposits and eradicates rust stains. It is perfect for use on tubs, shower heads, glassware, sinks, fiberglass, humidifiers, dishwashers and more. USDA Biopreferred Program Certified remover comes in a convenient bucket with a wire handle.