Learn More
Enterprise Technology Solutions SKILCRAFT Disinfectant,Broad-Spectrum,RTU,1 Gallon,4/BX,CL
SDP
Catalog No. 5000829977
Encompass_Preferred
PURE Hard Surface Disinfectant is an EPA-registered food-contact surface sanitizer and disinfectant that meets EPA criteria for use against SARS-CoV 2. It kills germs in as little as 30 seconds and offers 24-hour residual protection on hard, nonporous surfaces. This ready-to-use, odorless, noncaustic, non-irritating formula contains no VOCs and delivers board-spectrum power to eliminate dangerous pathogens in a wide range of environments. Use on painted, glazed tile, plastic, metal, glass and glazed porcelain in homes, offices, restaurants, schools, hotels, restrooms, recreational facilities and public transport vehicles.
Catalog No. 50-008-29977 Supplier Enterprise Technology Solutions Supplier No. NSN6911873
PURE Hard Surface Disinfectant is an EPA-registered food-contact surface sanitizer and disinfectant that meets EPA criteria for use against SARS-CoV 2. It kills germs in as little as 30 seconds and offers 24-hour residual protection on hard, nonporous surfaces. This ready-to-use, odorless, noncaustic, non-irritating formula contains no VOCs and delivers board-spectrum power to eliminate dangerous pathogens in a wide range of environments. Use on painted, glazed tile, plastic, metal, glass and glazed porcelain in homes, offices, restaurants, schools, hotels, restrooms, recreational facilities and public transport vehicles.