Learn More
Enterprise Technology Solutions SKILCRAFT Exam Glove,Powder-Free,Small,100/BX,10/CT,Blue
SDP
Catalog No. 5000828305
Encompass_Preferred
Ambidextrous, medical-grade powder-free gloves are made of 100 percent synthetic nitrile polymer and offer a solution to those allergic to natural latex protein. Tested for use with chemotherapy drugs, ambidextrous gloves are puncture-resistant and chemical-resistant. These 9.5" long nonsterile gloves are 4 mil thick. Textured fingertips provide a strong grip for both wet and dry applications.
Catalog No. 50-008-28305 Supplier Enterprise Technology Solutions Supplier No. NSNNIB0236
Ambidextrous, medical-grade powder-free gloves are made of 100 percent synthetic nitrile polymer and offer a solution to those allergic to natural latex protein. Tested for use with chemotherapy drugs, ambidextrous gloves are puncture-resistant and chemical-resistant. These 9.5" long nonsterile gloves are 4 mil thick. Textured fingertips provide a strong grip for both wet and dry applications.