Learn More
Enterprise Technology Solutions SKILCRAFT Exam Gloves, Vinyl, Large, 100/BX, Opaque Blue
SDP
Catalog No. 5000827801
Encompass_Preferred
Powder-free vinyl exam gloves are ideal for package handling and airport screeners. Ambidextrous, medical-grade gloves are made of 100 percent synthetic vinyl for users who are sensitive to natural rubber protein. These nonsterile gloves are 4 mil thick.
Catalog No. 50-008-27801 Supplier Enterprise Technology Solutions Supplier No. NSNNIB0106
Powder-free vinyl exam gloves are ideal for package handling and airport screeners. Ambidextrous, medical-grade gloves are made of 100 percent synthetic vinyl for users who are sensitive to natural rubber protein. These nonsterile gloves are 4 mil thick.