Learn More
Enterprise Technology Solutions SKILCRAFT Floor Pads,Polishing,3400Series,11/16" thick,17"Dia,5/BX,Tan
SDP
Catalog No. 5000790327
Encompass_Preferred
Made of 3M floor pad materials, 11/16" thick tan burnishing pad is used for rotary disk floor machines above 1500 rpm. Use on resilient tile, terrazzo, marble and ceramic floors. Corrosion-resistant pads can be washed and reused. They produce a "wet look" gloss in high-traffic areas.
Catalog No. 50-007-90327 Supplier Enterprise Technology Solutions Supplier No. NSN5132230
Made of 3M floor pad materials, 11/16" thick tan burnishing pad is used for rotary disk floor machines above 1500 rpm. Use on resilient tile, terrazzo, marble and ceramic floors. Corrosion-resistant pads can be washed and reused. They produce a "wet look" gloss in high-traffic areas.