Learn More
Enterprise Technology Solutions SKILCRAFT Floor Pads,Polishing,4100 Series,20",5/BX,White
SDP
Catalog No. 5000790245
Encompass_Preferred
Made of 3M floor pad materials, this polishing floor pad delivers long-lasting performance. Use this white polishing pad to buff very soft finishes or polish soft waxes on wood floors. It is designed for 175-600 rpm.
Catalog No. 50-007-90245 Supplier Enterprise Technology Solutions Supplier No. NSN5017027
Made of 3M floor pad materials, this polishing floor pad delivers long-lasting performance. Use this white polishing pad to buff very soft finishes or polish soft waxes on wood floors. It is designed for 175-600 rpm.