Learn More
Enterprise Technology Solutions SKILCRAFT Gloves,Exam,Nitrile,PF,9.5",Large,100/Box,10/CT,Blue
SDP
Catalog No. 5000828308
Encompass_Preferred
Medical-Grade Synthetic Gloves offer a solution to those allergic to natural latex protein. 100 percent synthetic nitrile polymer is nonsterile, 4 mil thick, powder-free and textured on the fingertips for a strong grip in both wet and dry applications. It resists punctures and chemicals. Ambidextrous, 9.5" long gloves are not made with natural rubber latex. They are tested for use with chemotherapy drugs and mandatory only for U.S. Department of Veterans Affairs.
Catalog No. 50-008-28308 Supplier Enterprise Technology Solutions Supplier No. NSNNIB0238
Medical-Grade Synthetic Gloves offer a solution to those allergic to natural latex protein. 100 percent synthetic nitrile polymer is nonsterile, 4 mil thick, powder-free and textured on the fingertips for a strong grip in both wet and dry applications. It resists punctures and chemicals. Ambidextrous, 9.5" long gloves are not made with natural rubber latex. They are tested for use with chemotherapy drugs and mandatory only for U.S. Department of Veterans Affairs.