Learn More
Enterprise Technology Solutions SKILCRAFT Multipurpose Cleaner, Biobased, 1 Gal, 4/BX, Citrus Scent/YW
SDP
Catalog No. 5000829839
Encompass_Preferred
Multipurpose cleaner handles everyday cleaning tasks as well as heavier-duty challenges. Multipurpose cleaner cleans a variety of soils and works on a variety of surfaces such as glass, mirrors, floors, walls, restroom and kitchen surfaces, and tile - almost any surface not harmed by water. Apply with a mop, brush, pressure washer, auto scrubber or use a spray and wipe application. TriBase Multipurpose Cleaner contains a 72 percent bio-based formula that contains three main bio-based materials and low VOCs and is phosphate-free. The three main surfactant ingredients are derived from corn, coconut and palm kernel, each supporting a different area of agriculture. Multipurpose cleaner is golden yellow in color and has a natural citrus fragrance. pH ranges from 8.0 to 9.0. Dilution ratios include 1:128 to 1:40 for light-duty cleaning and 1:32 to 1:10 for medium/heavy-duty cleaning/degreasing.
Catalog No. 50-008-29839 Supplier Enterprise Technology Solutions Supplier No. NSN5552901
Multipurpose cleaner handles everyday cleaning tasks as well as heavier-duty challenges. Multipurpose cleaner cleans a variety of soils and works on a variety of surfaces such as glass, mirrors, floors, walls, restroom and kitchen surfaces, and tile - almost any surface not harmed by water. Apply with a mop, brush, pressure washer, auto scrubber or use a spray and wipe application. TriBase Multipurpose Cleaner contains a 72 percent bio-based formula that contains three main bio-based materials and low VOCs and is phosphate-free. The three main surfactant ingredients are derived from corn, coconut and palm kernel, each supporting a different area of agriculture. Multipurpose cleaner is golden yellow in color and has a natural citrus fragrance. pH ranges from 8.0 to 9.0. Dilution ratios include 1:128 to 1:40 for light-duty cleaning and 1:32 to 1:10 for medium/heavy-duty cleaning/degreasing.