Learn More
Enterprise Technology Solutions SKILCRAFT Recordable DVD, Encrypted, 4.7GB, 120min, 25/Spindle, Silver
SDP
Supplier:  Enterprise Technology Solutions NSN5992658
Encompass_Preferred
DVD media offers built-in burning and encryption features. Exclusive, EncryptDisc built-in disc burning does not allow unencrypted data to be written to the disc. Each disc contains Federal Information Processing Standard certified 256-bit AES Encryption, 140-2, Level 1 (Certificates 562/819). Use recordable DVDs on a spindle for data archiving, data distribution, data storage and more on Windows 2000, XP, Vista and Windows 7. Each encrypted, FIPS 140-2 certified disc holds up to 4.7GB and 120 minutes and offers an 8X record speed.
Catalog No. 50-007-40432