Learn More
Enterprise Technology Solutions SKILCRAFT Safety Marking Tape, 2"x108', Yellow
SDP
Catalog No. 5000828323
Encompass_Preferred
Floor-safety marking tape is made of heavy-duty poly vinyl with rubber adhesive that sticks tight and removes cleanly. Tape resists abrasion, moisture, common solvents and weather. Ideal for indoor or outdoor marking of traffic areas, emergency zones and work sites. Bright yellow color makes it perfect for color-coding (yellow for gases). Flexible backing allows application to curved walls, ducts, low-hanging objects as well as floors. 3" core is made of 100 percent post-consumer recycled content. 2" wide x 108" long roll of tape meets OSHA color-coding requirements and conforms to ASTM-D 3330, 3759, 3652 standards.
Catalog No. 50-008-28323 Supplier Enterprise Technology Solutions Supplier No. NSN6174257
Floor-safety marking tape is made of heavy-duty poly vinyl with rubber adhesive that sticks tight and removes cleanly. Tape resists abrasion, moisture, common solvents and weather. Ideal for indoor or outdoor marking of traffic areas, emergency zones and work sites. Bright yellow color makes it perfect for color-coding (yellow for gases). Flexible backing allows application to curved walls, ducts, low-hanging objects as well as floors. 3" core is made of 100 percent post-consumer recycled content. 2" wide x 108" long roll of tape meets OSHA color-coding requirements and conforms to ASTM-D 3330, 3759, 3652 standards.