Learn More
Learning Resources Skill Builders Summer Learning Activity Set for Pre-K to K

Description
- Get kids ready for kindergarten with comprehensive learning activities that promote active, outdoor play
- Kit includes activities to help children learn letter, number, and color recognition, count to 100, learn to read and write, etc.
- Activities including alphabet yoga, reading games, and action-packed movement prompts get kids outside and moving as they learn.
- Comes with a fun write-and-wipe activity book filled with reading games and handwriting practice for kids, plus a summer calendar to help them stay on track and count down the days until school starts.
- Includes a reusable storage box for easy clean-up
- Made from durable, BPA-free plastic
Specifications
Specifications
| Grade Level | Early Childhood, Elementary |
| Grade | Pre-K to K |
| Class Size | 1 Class |
| Product Type | Summer Learning Activity Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.