Learn More
Tosoh Bioscience LLC SkillPak TSKgel SuperQ-5PW (20) 5 mL column
Catalog No. NC1899308 Shop All Tosoh Bioscience LLC Products
Encompass
The SkillPak TSKgel SuperQ-5PW (20) 5 mL column is designed for fast method development, resin screening or sample concentration of biomolecules.
Catalog No. NC1899308 Supplier Tosoh Bioscience LLC Supplier No. 0045244
The SkillPak TSKgel SuperQ-5PW (20) 5 mL column is designed for fast method development, resin screening or sample concentration of biomolecules.