Learn More
MOLTOX Molecular Toxicology Inc 15% Skim Milk, 50 mL/glass bottle, 1 bottle/pack, Store Refrigerated
SDP
Catalog No. 50948670
Encompass_Preferred
Skim Milk can be used for maintaining frozen stock cultures as well as for the cultivation and enumeration of many common microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-670 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S50
Skim Milk can be used for maintaining frozen stock cultures as well as for the cultivation and enumeration of many common microorganisms. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.