Learn More
NEOGEN Skim Milk, Agglomerated NFDM, 5 kg
SDP
Supplier:  NEOGEN 700003502
Encompass_Preferred
Skim Milk, Agglomerated NFDM is dehydrated skim milk for use in preparing microbiological culture media in a laboratory setting. Skim Milk, Agglomerated NFDM is not intended for use in the diagnosis of disease or other conditions in humans. Skim Milk, Agglomerated NFDM is soluble, spray-dried skim milk. When prepared in a 10% solution, it is equivalent to fresh skim milk. Skim Milk is added to enhance growth of Mycobacterium tuberculosis and Corynebacterium diphtheria in culture media. Skim Milk, Agglomerated NFDM is used for differentiating organisms based on coagulation and proteolysis of casein in foods and dairy products. Indicators of pH, Litmus and Bromocresol Purple, are used with Skim Milk, Agglomerated NFDM to determine acid production. Methylene Blue and Resazurin may be used with Skim Milk, Agglomerated NFDM as oxidation-reduction indicators.For more information visit https://www.neogen.com/solutions/Microbiology/skim-milk-agglomerated-nfdm
Catalog No. 50-201-5736