Learn More
Key Scientific Products CryoCare, Organism Preservative System, Skim Milk Base in Broth, 50 Vials per Box
SDP
Catalog No. NC0185112
Encompass_Preferred
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains skim milk base in a broth. Includes 50 vials per box.
Catalog No. NC0185112 Supplier Key Scientific Products Supplier No. KSMB50
CryoCare organism preservative system lowers the cost of stock cultures by stretching each expensive culture into 15 or more which meet NCCLS standards. Vial contains skim milk base in a broth. Includes 50 vials per box.