Learn More
NEOGEN Skim Milk Powder, 10 kg
SDP
Catalog No. 502015854
Encompass_Preferred
Skim Milk Powder is used when preparing microbiology culture media in a laboratory setting. Milk Powder is not intended for use in the diagnosis of disease or other conditions in humans. A bacteriological grade of thermophile free spray dried skim milk. Used in Milk Plate Count Agar NCM0119 and in media for diagnostic tests involving the digestion or coagulation of casein and the fermentation of lactose. Recommended working concentration 10%.For more information visit https://www.neogen.com/solutions/Microbiology/skim-milk-powder
Catalog No. 50-201-5854 Supplier NEOGEN Supplier No. 700004714
Skim Milk Powder is used when preparing microbiology culture media in a laboratory setting. Milk Powder is not intended for use in the diagnosis of disease or other conditions in humans. A bacteriological grade of thermophile free spray dried skim milk. Used in Milk Plate Count Agar NCM0119 and in media for diagnostic tests involving the digestion or coagulation of casein and the fermentation of lactose. Recommended working concentration 10%.For more information visit https://www.neogen.com/solutions/Microbiology/skim-milk-powder