Learn More
Bioworld NFDM (Non-Fat Dry Milk Powder) / Skim Milk Powder 1kg
Catalog No. 50254369
Encompass_Preferred
Used as a complete medium, or incorporated into other media, for isolation and cultivation of microorganisms found in milk products and to demonstrate coagulation and proteolysis of casein. For Western Blots (18 months shelf life) order product number 30620122 which contains Sodium Azide and Ampicillin to prevent bacterial growth. Appearance Form : Powder, Appearance Color : Yellow, 1kg
Catalog No. 50-254-369 Supplier Bioworld Supplier No. 306200742
Used as a complete medium, or incorporated into other media, for isolation and cultivation of microorganisms found in milk products and to demonstrate coagulation and proteolysis of casein. For Western Blots (18 months shelf life) order product number 30620122 which contains Sodium Azide and Ampicillin to prevent bacterial growth. Appearance Form : Powder, Appearance Color : Yellow, 1kg