Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Skim Milk Powder

Catalog No. MP290288705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290288705 5 lb.
MP290288725 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP290288705 Supplier MP Biomedicals™ Supplier No. 0290288705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A nonfat dry milk powder is produced from fresh skim milk. Skim Milk Powder is a component used in formulating animal research diets.

  • Skim milk powder
  • Used in formulating research animal diets
  • It is heated to achieve denaturation of the whey proteins
  • Proteins, enzymes & peptides
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Formulating research animal diet
Quantity 5 lb.
Storage Requirements Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.