Learn More
Bioworld NFDM/Skim Milk Powder, 500 g
Supplier:  Bioworld 306200741
Encompass_Preferred
Skim Milk PowderUsed as a complete medium, or incorporated into other media, for isolation and cultivation of microorganisms found in milk products and to demostrate coagulation and proteolysis of caseinFor Western Blots (18 months shelf life) order product number 30620122which contains Sodium Azide and Ampicillin to prevent bacterial growth
Catalog No. 50-254-368