Learn More
Bioworld Skim Milk Powder with Sodium Azide and Ampicillin, 500g
Catalog No. NC2057745
Encompass_Preferred
Skim Milk powder. - Used in Western Blots - Contains Sodium Azide and Ampicillin to prevent bacterial growth.
Catalog No. NC2057745 Supplier Bioworld Supplier No. 306201221
Skim Milk powder. - Used in Western Blots - Contains Sodium Azide and Ampicillin to prevent bacterial growth.