Learn More
NEOGEN Skim Milk Powder, 500 g
SDP
Catalog No. 502015852
Encompass_Preferred
This item has a minimum order quantity of 5 per supplier requirements
Skim Milk Powder is used when preparing microbiology culture media in a laboratory setting. Milk Powder is not intended for use in the diagnosis of disease or other conditions in humans. A bacteriological grade of thermophile free spray dried skim milk. Used in Milk Plate Count Agar NCM0119 and in media for diagnostic tests involving the digestion or coagulation of casein and the fermentation of lactose. Recommended working concentration 10%.For more information visit https://www.neogen.com/solutions/Microbiology/skim-milk-powder
Catalog No. 50-201-5852 Supplier NEOGEN Supplier No. 700004712
This item has a minimum order quantity of 5 per supplier requirements
Skim Milk Powder is used when preparing microbiology culture media in a laboratory setting. Milk Powder is not intended for use in the diagnosis of disease or other conditions in humans. A bacteriological grade of thermophile free spray dried skim milk. Used in Milk Plate Count Agar NCM0119 and in media for diagnostic tests involving the digestion or coagulation of casein and the fermentation of lactose. Recommended working concentration 10%.For more information visit https://www.neogen.com/solutions/Microbiology/skim-milk-powder