Learn More
Thermo Scientific 2.8 mm (High Sensitivity) Skimmer Cone Insert
Supplier:  Thermo Scientific 1311880
Encompass
2.8 mm Skimmer Cone insert for iCAP Q/Qnova ICP-MS Systems – High Sensitivity Applications
Catalog No. NC0740016