Learn More
Glass Expansion Skimmer ConeGuard
SDP
Supplier:  Glass Expansion 708031008
Encompass_Preferred
Skimmer ConeGuard. For use with Agilent 7700x/7800/7850/8800/8900 ICP-MS skimmer cones to protect cone threads during cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3073