Learn More
Glass Expansion Skimmer ConeGuard PerkinElmer Elan Series
SDP
Catalog No. 502703072
Encompass_Preferred
Skimmer ConeGuard PerkinElmer Elan Series. For use with PerkinElmer 6000/9000/DRCII/DRC-E ICP-MS skimmer cones to protect cone threads during cleaning. Compatible with Nickel Skimmer Cone NC1585740 and Platinum Skimmer Cone 501907683. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3072 Supplier Glass Expansion Supplier No. 708031022
Skimmer ConeGuard PerkinElmer Elan Series. For use with PerkinElmer 6000/9000/DRCII/DRC-E ICP-MS skimmer cones to protect cone threads during cleaning. Compatible with Nickel Skimmer Cone NC1585740 and Platinum Skimmer Cone 501907683. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When cones are not in use, keep them stored in original packaging for protection.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001