Learn More
Cayman Chemical Skimmianine 10mg
SDP
Catalog No. 502950694
Encompass_Preferred
An internal standard for the quantification of sodium butyrate by GC- or LC-MS
Catalog No. 50-295-0694 Supplier Cayman Chemical Supplier No. 34643 10MG
An internal standard for the quantification of sodium butyrate by GC- or LC-MS