Learn More
Braintree Scientific Skin Button w/Dacron Patch, and Silicone Sleeve
SDP
Catalog No. 501955861
Encompass
Skin ButtonRodent Skin Buttons are used for externalizing a catheter or for an attachment of a spring tether to the animal as an alternative to the harness or jacket. The advantage of a skin button is lower chance of infection, secure catheter anchoring and the ability to keep the catheter away from the animal. The silicone sleeve is the perfect size for most all standard rodent tethers and the 1" Dacron Mesh allows for suture attachment and tissue ingrowth. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5861 Supplier Braintree Scientific Supplier No. SBD02
Skin ButtonRodent Skin Buttons are used for externalizing a catheter or for an attachment of a spring tether to the animal as an alternative to the harness or jacket. The advantage of a skin button is lower chance of infection, secure catheter anchoring and the ability to keep the catheter away from the animal. The silicone sleeve is the perfect size for most all standard rodent tethers and the 1" Dacron Mesh allows for suture attachment and tissue ingrowth. **Sold for research purposes only and not for human use**