Learn More
Braintree Scientific Skin Button w/Dacron Patch, for Mice
SDP
Catalog No. 501955862
Encompass
Skin ButtonMouseSkin Buttons are used for externalizing a catheter, secure catheter anchoring and the ability to keep the catheter away from the animal. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5862 Supplier Braintree Scientific Supplier No. SBDM2
Skin ButtonMouseSkin Buttons are used for externalizing a catheter, secure catheter anchoring and the ability to keep the catheter away from the animal. **Sold for research purposes only and not for human use**