Learn More
StatLab Skin Control Slides
SDP
Catalog No. 22024939
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22-024-939 25 Pk.
1 options
Catalog No. 22-024-939 Supplier StatLab Supplier No. CSS0825P
Add to Cart
Add to Cart
Use skin control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Product Type Control Slide
Quantity 25 Pk.