Description
- Pack of 10 sterile, high-precision blades
- Designed for smooth, consistent skin harvests
- Ultra-sharp stainless steel for clean, uniform graft thickness
- Compatible with standard Watson Dermatome Handles
- Individually packaged to maintain sterility and safety
- Ideal for burn care, reconstructive surgery, specimen harvesting, and autopsy/postmortem graft collection
- Precision-engineered handle designed for controlled, smooth skin grafting
- Compatible with standard dermatome/grafting blades
- Textured grip enhances stability during delicate cutting
- Rigid stainless-steel construction ensures long-term durability
- Fully autoclavable and corrosion-resistant
- Suitable for surgical grafting, research labs, burn units, and postmortem tissue procurement
Specifications
Specifications
| Blade Material | Stainless Steel |
| Product Type | Handles |
| For Use With (Equipment) | Skin Grafting Blades |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Certifications/Compliance | ISO/GMP |
| Autoclavable | Autoclavable |
| Handle Material | Stainless Steel |
| Graduated | No |