Learn More
ATCC Skin Microbiome; Whole Cell Mix
Catalog No. 502383124
Encompass
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product is prepared as an even mixture of whole cells and comprises fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical skin microbial communities. Each order for this product is provided with access to run 10 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 1.2x108 cells/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only
Catalog No. 50-238-3124 Supplier ATCC Supplier No. MSA2005
 Restricted Item
Add to Cart
Add to Cart
NGS Standards are mock microbial communities that mimic mixed metagenomic samples. This product is prepared as an even mixture of whole cells and comprises fully sequenced, characterized, and authenticated bacterial species observed in normal and atypical skin microbial communities. Each order for this product is provided with access to run 10 analyses on One Codex, the leading bioinformatics platform for microbial genomics and metagenomics. Specification Range: 1.2x108 cells/vial ± 1 log* *Whole cell concentrations indicate ATCC manufacturing specifications and are provided as a reference only