Learn More
Med Vet International 3M Duraprep Skin Prep Solution, 26 mL, Foam Applicator, 0.7%/74% Strength, Iodine Povacrylex/Isopropyl Alcohol, Nonsterile, 8630, 20/Case
Catalog No. 502829676
Encompass_Preferred
3M™ Duraprep™ surgical solution comes in a convenient applicator for quick, efficient skin coverage It forms a thick layer of film that quickly kills bacteria and continues to prevent microbial build-up When dry, it stays in place even during operations and other procedures This film serves as a primer that holds incise drapes securely in place Duraprep™ 26 milliliter reduces the chance of infection The film is easy to remove using saline or a sponge
Catalog No. 50-282-9676 Supplier Med Vet International Supplier No. 172618CS
3M™ Duraprep™ surgical solution comes in a convenient applicator for quick, efficient skin coverage It forms a thick layer of film that quickly kills bacteria and continues to prevent microbial build-up When dry, it stays in place even during operations and other procedures This film serves as a primer that holds incise drapes securely in place Duraprep™ 26 milliliter reduces the chance of infection The film is easy to remove using saline or a sponge