Learn More
STERILAB INC SKIN PROCESSING 7X18IN PK25
Catalog No. 501317560
Encompass_Preferred
Skin processing boards 7"x18" ALL-POLY LABboards designed specifically for processing dematome recovered skin for allografts. Printed with metric perimeter rulers and centimeter field grid. 25-count pack.. Disposable.
Catalog No. 50-131-7560 Supplier STERILAB INC Supplier No. 0718A
Skin processing boards 7"x18" ALL-POLY LABboards designed specifically for processing dematome recovered skin for allografts. Printed with metric perimeter rulers and centimeter field grid. 25-count pack.. Disposable.