Learn More
Med Vet International Skin Protectant 3M Cavilon No Sting 28 mL Spray Bottle Liquid Chg Compatible, 3346, 12/Case
Catalog No. 502831963
Encompass_Preferred
This 3M™ No Sting film spray protects skin from damage resulting from bodily fluids, adhesives or friction It contains no alcohol and is hypoallergenic to prevent discomfort It dries rapidly and is non-sticky, waterproof and breathable for comfort It gives you a long-lasting skin barrier that's suitable for individuals with incontinence-related skin issues or a stoma
Catalog No. 50-283-1963 Supplier Med Vet International Supplier No. 281799CS
This 3M™ No Sting film spray protects skin from damage resulting from bodily fluids, adhesives or friction It contains no alcohol and is hypoallergenic to prevent discomfort It dries rapidly and is non-sticky, waterproof and breathable for comfort It gives you a long-lasting skin barrier that's suitable for individuals with incontinence-related skin issues or a stoma