Learn More
Med Vet International Skin Barrier Applicator 3M Cavilon No Sting 26 To 62% Strength Hexamethyldisiloxane / Isooctane / Acrylate Terpolymer / Polyphenylmethylsiloxane Individual Packet Sterile, 3mL, 3345, 100/Case
Catalog No. 502830158
Encompass_Preferred
3M™ Cavilon™ no sting barrier film protects the skin from body fluids and friction It has an alcohol-free formula that is gentle on the skin It is breathable and dries quickly without leaving a residue for enhanced comfort The barrier film is flexible, enabling it to contour around bends and creases It is non-cytotoxic and hypoallergenic to minimize irritations
Catalog No. 50-283-0158 Supplier Med Vet International Supplier No. 292019CS
3M™ Cavilon™ no sting barrier film protects the skin from body fluids and friction It has an alcohol-free formula that is gentle on the skin It is breathable and dries quickly without leaving a residue for enhanced comfort The barrier film is flexible, enabling it to contour around bends and creases It is non-cytotoxic and hypoallergenic to minimize irritations