Learn More
Med Vet International Skin Barrier Applicator 3M Cavilon No Sting 26 To 62% Strength Hexamethyldisiloxane / Isooctane / Acrylate Terpolymer / Polyphenylmethylsiloxane Individual Packet Sterile, 1mL, 3343, 25/Box
Catalog No. 502830154
Encompass_Preferred
The 3M™ skin barrier protects both intact and damaged skin from friction, incontinence and adhesive trauma The formula is hypoallergenic and latex- and alcohol-free, so it won't irritate or sting skin when applied It's non-sticky, waterproof and dries quickly to enhance comfort The flexible Cavilon™ barrier film conforms to the skin to guarantee protection The no-sting barrier film is CHG compatible, so it won't interfere with the effectiveness of antibacterial treatments
Catalog No. 50-283-0154 Supplier Med Vet International Supplier No. 292018BX
The 3M™ skin barrier protects both intact and damaged skin from friction, incontinence and adhesive trauma The formula is hypoallergenic and latex- and alcohol-free, so it won't irritate or sting skin when applied It's non-sticky, waterproof and dries quickly to enhance comfort The flexible Cavilon™ barrier film conforms to the skin to guarantee protection The no-sting barrier film is CHG compatible, so it won't interfere with the effectiveness of antibacterial treatments