Learn More
Surgical Design™ Skin Punch Plier
Catalog No. 21124609 Shop All Surgical Design Products
Change view
Click to view available options
Size:
Various
Catalog No. Size
21-124-609 Various
1 options
Catalog No. 21-124-609 Supplier Surgical Design™ Supplier No. 54786
Add to Cart
Add to Cart
Skin punch plier delivers clean, precise skin biopsies and tissue sampling, with stainless-steel durability and controlled handling for consistent results.
  • Constructed from premium surgical-grade stainless steel for precision, strength, and durability
  • Designed to produce clean, controlled tissue punches with minimal trauma
  • Ergonomic plier-style handle ensures stability and comfort for the operator
  • Sharp, fine cutting edge delivers accurate circular excisions for biopsy or graft preparation
  • Integrated adjustment screw allows controlled depth and consistent results
  • Smooth spring action reduces hand fatigue during repetitive use
  • Autoclavable and reusable, maintaining consistent cutting performance
  • Ideal for skin biopsies, graft preparation, and tissue sampling
  • Suitable for dermatology, pathology, and cosmetic surgical environments
Specifications
Product Type Dermal Punch
Size Various
Disposable Reusable
Sterility Non-sterile
Autoclavable Autoclavable
Graduated No