Learn More
Med Vet International Oasis Surgical Staple Remover, Sterile, Disposable and Single-Use, Plastic Handle, Each
Catalog No. 502522167
Encompass_Preferred
Versatile single-use skin stapler allows for fast and easy removal of surgical skin staples with minimal discomfort to the patient. Consistent performance by placing the metal tip of the sterile remover underneath the staple and squeezing the plastic handles until they touch and the extractor is fully closed. Once the staple is completely reformed the remover can be lifted from the skin and the staple is removed. Each sterile, single-use instrument is shipped ready-to-use in sealed plastic packaging that's easy to open. Surgical Staplers and surgical staple removers are perfect for wound closure Sold in eaches.
Catalog No. 50-252-2167 Supplier Med Vet International Supplier No. STAPLEREMPL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Versatile single-use skin stapler allows for fast and easy removal of surgical skin staples with minimal discomfort to the patient. Consistent performance by placing the metal tip of the sterile remover underneath the staple and squeezing the plastic handles until they touch and the extractor is fully closed. Once the staple is completely reformed the remover can be lifted from the skin and the staple is removed. Each sterile, single-use instrument is shipped ready-to-use in sealed plastic packaging that's easy to open. Surgical Staplers and surgical staple removers are perfect for wound closure Sold in eaches.