Learn More
Med Vet International AHS Skin Stapler, Sterile, 35 Regular, AHS35 Regular Each
Supplier:  Med Vet International AHS35R
Encompass_Preferred
AHS Skin Stapler is a top quality, sterile, single use, skin staple. This stapler comes with (35) regular medical grade stainless steel staples and measure 5.5mm in length, 4mm wide, and has a diameter of .6mm. This staper features: A unique ratchet system to prevent jamming Ergonomic, user friendly design Easy to check remaining staples in cartridge Clear view of operating site Short trigger stroke for ease of use and nonstop feed Angled head allows visibility of incision to ensure precise staple placement Alternative to products like ZipStitch instant stitches and Sutures Easy to use For regular wounds Faster than Stitches Easier to remove than Stitches Fewer issues with scarring
Catalog No. 50-252-2461