Learn More
Med Vet International Sterile Surgical Skin Stapler, 35 Regular, Each
Catalog No. 502522543
Encompass_Preferred
The Oasis sterile skin stapler features 35 regular skin staples in a compact and versatile design. This surgical stapler’s easy operation allows for the accurate application of the medical staples for optimal performance. The quality construction of the Oasis sterile skin stapler provides consistent and reliable performance every time. Distinctive medical staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Regular surgical staples measure 5.4mm x 3.6mm
Catalog No. 50-252-2543 Supplier Med Vet International Supplier No. 35R
The Oasis sterile skin stapler features 35 regular skin staples in a compact and versatile design. This surgical stapler’s easy operation allows for the accurate application of the medical staples for optimal performance. The quality construction of the Oasis sterile skin stapler provides consistent and reliable performance every time. Distinctive medical staple shuttling feature greatly reduces track friction, a primary cause of jamming Short trigger stroke for ease of use and nonstop feed Handle design comfortably fits a wide range of hand sizes Angled head allows visibility of incision to ensure precise staple placement during operation Arrow indicator assists in accurate staple alignment Staple count indicator allows you to view staples remaining to help minimize waste Quality construction and simple design assure reliable consistent performance Regular surgical staples measure 5.4mm x 3.6mm